Learn More
Ran Biotechnologies Inc 50g of FO-101 fluorinated oil
SDP
Catalog No. NC3660272
Encompass_Preferred
50g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)
Catalog No. NC3660272 Supplier Ran Biotechnologies Inc Supplier No. FO10150G
50g of FO-101 fluorinated oil (bp 130degC; d1.55; viscosity 0.80cSt)