Learn More
Ran Biotechnologies Inc 10kg of FO-111 fluorinated oil
SDP
Catalog No. NC3660239
Encompass_Preferred
10kg of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660239 Supplier Ran Biotechnologies Inc Supplier No. FO11110000G
Add to Cart
Add to Cart
Request a Quote
10kg of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)