Learn More
Ran Biotechnologies Inc 100g of FO-111 fluorinated oil
SDP
Catalog No. NC3660297
Encompass_Preferred
100g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660297 Supplier Ran Biotechnologies Inc Supplier No. FO111100G
100g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)