Learn More
Ran Biotechnologies Inc 1.5kg of FO-111 fluorinated oil
SDP
Catalog No. NC3660236
Encompass_Preferred
1.5kg of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660236 Supplier Ran Biotechnologies Inc Supplier No. FO1111500G
1.5kg of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)