Learn More
Ran Biotechnologies Inc 200g of FO-111 fluorinated oil
SDP
Catalog No. NC3660298
Encompass_Preferred
200g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660298 Supplier Ran Biotechnologies Inc Supplier No. FO111200G
200g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)