Learn More
Ran Biotechnologies Inc 2.5kg of FO-111 fluorinated oil
SDP
Catalog No. NC3660238
Encompass_Preferred
2.5kg of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660238 Supplier Ran Biotechnologies Inc Supplier No. FO1112500G
2.5kg of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)