Learn More
Ran Biotechnologies Inc 300g of FO-111 fluorinated oil
SDP
Catalog No. NC3660299
Encompass_Preferred
300g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660299 Supplier Ran Biotechnologies Inc Supplier No. FO111300G
300g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)