Learn More
Ran Biotechnologies Inc 500g of FO-111 fluorinated oil
SDP
Catalog No. NC3660305
Encompass_Preferred
500g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660305 Supplier Ran Biotechnologies Inc Supplier No. FO111500G
500g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)