Learn More
Ran Biotechnologies Inc 50g of FO-111 fluorinated oil
SDP
Catalog No. NC3660295
Encompass_Preferred
50g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)
Catalog No. NC3660295 Supplier Ran Biotechnologies Inc Supplier No. FO11150G
50g of FO-111 fluorinated oil (bp 162degC; d1.606; viscosity 0.93cSt)