Learn More
AdipoGen FOAA
SDP
Supplier:  AdipoGen CDXF0022M250
Encompass_Preferred
Chemical. CAS 233759-98-3. Formula C22H14O7. MW 390.34. Synthetic. Fluorescent label which reacts with amines after activation to give isomerically homogeneous derivatives.
Catalog No. 50-256-0934