Learn More
CHEMetrics™ by AquaPhoenix FoalWatch™ Foaling Prediction Kit
SDP
Catalog No. 13299673
Change view
Click to view available options
No. of Tests:
20
Catalog No. No. of Tests
13-299-673 20
1 options
Catalog No. 13-299-673 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K1700
Add to Cart
Add to Cart
Foaling prediction test kit utilizing Titrets ampoules and reverse titration technology. Sample is drawn into the ampoule until a color change signals the endpoint.
  • FoalWatch offers a reliable, chemistry‐based way to anticipate foaling with confidence.
  • Designed to be gentle for both mare and handler, the kit requires no special training and includes everything needed to perform 20 tests
Specifications
No. of Tests 20
Type Foaling Prediction Kit