Learn More
Med Vet International McKesson Lite Silicone Gel Adhesive without Border Thin Silicone Foam Dressing, 6 x 6 Inch, 4894, Each
Catalog No. 503043827
Encompass_Preferred
Thin Silicone Foam Dressing McKesson Lite 6 X 6 Inch Square Silicone Gel Adhesive without Border Sterile The silicone gel adhesive holds securely in place and removes gently with minimal skin disturbance The gel sticks to healthy, dry skin and not the wound bed for added safety The dressings form a seal that prevents leaks The outer film layer is breathable for airflow while resisting moisture and bacteria An inner foam pad is highly absorbent The silicone gel adhesive dressings are repositionable and remain adherent Wounds stay moist for faster healing They are 6 x 6 inches and do not have a border
Catalog No. 50-304-3827 Supplier Med Vet International Supplier No. 1083094EA
Thin Silicone Foam Dressing McKesson Lite 6 X 6 Inch Square Silicone Gel Adhesive without Border Sterile The silicone gel adhesive holds securely in place and removes gently with minimal skin disturbance The gel sticks to healthy, dry skin and not the wound bed for added safety The dressings form a seal that prevents leaks The outer film layer is breathable for airflow while resisting moisture and bacteria An inner foam pad is highly absorbent The silicone gel adhesive dressings are repositionable and remain adherent Wounds stay moist for faster healing They are 6 x 6 inches and do not have a border