Learn More
Med Vet International McKesson Silicone Adhesive with Border Silicone Foam Dressing, 4 x 4 Inch, 4843, Each
Catalog No. 503044076
Encompass_Preferred
Silicone Foam Dressing McKesson 4 X 4 Inch Square Silicone Adhesive with Border Sterile Designed to affix to dry, intact skin to maintain a moist wound environment and promote healing Outer film layer is breathable while remaining resistant to bacteria and moisture infiltration Inner hydrocellular foam pad is absorbent Seals the wound margins to help prevent exudate from leaking on surrounding tissue Highly conformable to fit on body's contours Perforated silicone gel adhesive minimizes pain and trauma to wounds upon removal Dressing can be lifted and repositioned without losing its adherent properties Sterile dressing is intended for single-use applications 3 x 3 inch pad
Catalog No. 50-304-4076 Supplier Med Vet International Supplier No. 886432EA
Silicone Foam Dressing McKesson 4 X 4 Inch Square Silicone Adhesive with Border Sterile Designed to affix to dry, intact skin to maintain a moist wound environment and promote healing Outer film layer is breathable while remaining resistant to bacteria and moisture infiltration Inner hydrocellular foam pad is absorbent Seals the wound margins to help prevent exudate from leaking on surrounding tissue Highly conformable to fit on body's contours Perforated silicone gel adhesive minimizes pain and trauma to wounds upon removal Dressing can be lifted and repositioned without losing its adherent properties Sterile dressing is intended for single-use applications 3 x 3 inch pad