Learn More
Plastilite Corp FOAM COOLER IN BOX 12/CS
Catalog No. NC9890298
Encompass_Preferred
Mailing and Shipping Containers, Insulated Containers. EPS Shipper. Foam Insulated Container. Foam Shipper. Insulated Shipper. Insulated Shipper Container. Temperature-controlled Package Shipper. 1.5” thick-wall foam cooler with white outer box at 32ECT strength with “PERISHABLE” and up arrows on two sides. Durable and highly insulating for your temperature sensitive products such as pharmaceutical and biological materials. Use with dry ice, gel ice packs, foam brick ice packs, and PCMs as refrigerant material. With proper amounts of refrigerant material this cooler can protect up to four days depending on ambient shipping temperatures. ABX5-C12 cooler in box, 1” thick. Inside Dimensions of 9.625"x7.75"x5.875” offering .25 cubic feet for product and refrigerant material. This EPS foam cooler is recyclable at more than 400 locations across North America.
Catalog No. NC9890298 Supplier Plastilite Corp Supplier No. ABX5C12
Mailing and Shipping Containers, Insulated Containers. EPS Shipper. Foam Insulated Container. Foam Shipper. Insulated Shipper. Insulated Shipper Container. Temperature-controlled Package Shipper. 1.5” thick-wall foam cooler with white outer box at 32ECT strength with “PERISHABLE” and up arrows on two sides. Durable and highly insulating for your temperature sensitive products such as pharmaceutical and biological materials. Use with dry ice, gel ice packs, foam brick ice packs, and PCMs as refrigerant material. With proper amounts of refrigerant material this cooler can protect up to four days depending on ambient shipping temperatures. ABX5-C12 cooler in box, 1” thick. Inside Dimensions of 9.625"x7.75"x5.875” offering .25 cubic feet for product and refrigerant material. This EPS foam cooler is recyclable at more than 400 locations across North America.