Learn More
Grainger 3M Double Coated Polyethylene Foam Tape, White, 1 in., 1 ea
Catalog No. NC0644731
Encompass_Preferred
3M™ Double Coated Polyethylene Foam Tape 4492W is a closed-cell, polyethylene foam carrier tape formulated for demanding indoor and outdoor mounting, bonding, and joining applications. This conformable tape is designed to fill gaps and distribute stress uniformly, making it suitable for bonding to rough or irregular surfaces.
  • Features high-strength acrylic adhesive for long-term durability
  • Provides high ultimate adhesion to a wide variety of surfaces including glass, metal, and various plastics
  • Conformable foam construction allows for bonding to irregular surfaces
  • Includes a white kraft paper release liner
  • Solvent-resistant with good initial tack
  • Suitable for applications such as decorative trim, nameplates, and automotive mirror bonding
Catalog No. NC0644731 Supplier Grainger Supplier No. 15C259
3M™ Double Coated Polyethylene Foam Tape 4492W is a closed-cell, polyethylene foam carrier tape formulated for demanding indoor and outdoor mounting, bonding, and joining applications. This conformable tape is designed to fill gaps and distribute stress uniformly, making it suitable for bonding to rough or irregular surfaces.
  • Features high-strength acrylic adhesive for long-term durability
  • Provides high ultimate adhesion to a wide variety of surfaces including glass, metal, and various plastics
  • Conformable foam construction allows for bonding to irregular surfaces
  • Includes a white kraft paper release liner
  • Solvent-resistant with good initial tack
  • Suitable for applications such as decorative trim, nameplates, and automotive mirror bonding