Learn More
Chemglass Life Sciences Foam Dewar Flask, 500mL, Low Form, Interior Dimensions: Depth x Dia. (mm): 70 x 114
SDP
Catalog No. 501531169
Encompass_Preferred
Foam Dewar Flask, 500mL, Low Form, Interior Dimensions: Depth x Dia. (mm): 70 x 114. Patented low form foam dewars provide a safe and durable vessel for handling liquid nitrogen, dry ice/acetone mix and cryogenic materials. Unlike conventional glass and Stainless Steel Dewar flasks which depend upon a vacuum layer for insulation, closed cell foam dewars use a fine closed cell foam for insulation. This foam material is chemically inert and mechanically durable enough to be exposed directly to liquid nitrogen, without leaking or degrading. Unique design makes the foam dewar easier to handle and safer to use than a traditional low profile glass Dewar. The lower thermal mass of the foam dewar will cause less liquid nitrogen boil off when it is filled. It also will accumulate less frost during regular use. The end result is that less liquid nitrogen is consumed.
Catalog No. 50-153-1169 Supplier Chemglass Life Sciences Supplier No. CG1590F500
Foam Dewar Flask, 500mL, Low Form, Interior Dimensions: Depth x Dia. (mm): 70 x 114. Patented low form foam dewars provide a safe and durable vessel for handling liquid nitrogen, dry ice/acetone mix and cryogenic materials. Unlike conventional glass and Stainless Steel Dewar flasks which depend upon a vacuum layer for insulation, closed cell foam dewars use a fine closed cell foam for insulation. This foam material is chemically inert and mechanically durable enough to be exposed directly to liquid nitrogen, without leaking or degrading. Unique design makes the foam dewar easier to handle and safer to use than a traditional low profile glass Dewar. The lower thermal mass of the foam dewar will cause less liquid nitrogen boil off when it is filled. It also will accumulate less frost during regular use. The end result is that less liquid nitrogen is consumed.