Learn More
Chemglass Life Sciences Foam Dewar Flask, 1, 400mL, Low Form, Interior Dimensions: Depth x Dia. (mm): 95 x 160
SDP
Catalog No. NC0881058
Encompass_Preferred
Foam Dewar Flask, 1,400mL, Low Form,Interior Dimensions: Depth x Dia. (mm): 95 x 160
Catalog No. NC0881058 Supplier Chemglass Life Sciences Supplier No. CG1590F1400
Foam Dewar Flask, 1,400mL, Low Form,Interior Dimensions: Depth x Dia. (mm): 95 x 160