Learn More
Grainger 3M, Double Sided Foam Tape, 3M
Supplier:  Grainger 15C395
Encompass_Preferred
The 3M Double Sided Foam Tape is a high-performance acrylic adhesive tape with a polyethylene foam carrier. It has excellent durability and bonding strength on a wide range of materials including metals, plastics, glass, and painted surfaces. This tape is designed for permanent bonding applications and can withstand temperature extremes, UV light exposure, and high humidity environments. It has quick stick at room temperature and high initial tack. This 3M tape comes in various thicknesses from 0.5mm to 3.2mm and widths from 6mm to 54mm. It can be die-cut for custom shapes. Overall, this is a versatile, heavy-duty foam tape ideal for permanent bonding in demanding applications.
Catalog No. NC1410890