Learn More
MSC PURELL Hand Sanitizer Refill, Ultra Nourishing Foam, 1200 mL
Catalog No. 502836921
Encompass
PURELL Advanced Hand Sanitizer ULTRA NOURISHING Foam is an alcohol-based formula designed to kill 99.99% of germs that may cause illness. This gentle foam formulation improves skin condition and is compatible with ES10 Touch-Free Hand Sanitizer Dispensers.
  • Gentle foam formulation.
  • Improves skin condition, clinically proven in as little as three days.
  • Enhanced with a moisturizer and antioxidant blend.
  • Kills 99.99% of germs that may cause illness.
  • Gentle on hands for everyday use.
  • Manufactured with high-quality ethyl alcohol and meets FDA safety standards.
  • Certified as ECOLOGO UL 2783 and USDA Biobased Product.
  • Compatible with ES10 automatic hand sanitizer dispensers.
Catalog No. 50-283-6921 Supplier MSC Supplier No. 59581744
PURELL Advanced Hand Sanitizer ULTRA NOURISHING Foam is an alcohol-based formula designed to kill 99.99% of germs that may cause illness. This gentle foam formulation improves skin condition and is compatible with ES10 Touch-Free Hand Sanitizer Dispensers.
  • Gentle foam formulation.
  • Improves skin condition, clinically proven in as little as three days.
  • Enhanced with a moisturizer and antioxidant blend.
  • Kills 99.99% of germs that may cause illness.
  • Gentle on hands for everyday use.
  • Manufactured with high-quality ethyl alcohol and meets FDA safety standards.
  • Certified as ECOLOGO UL 2783 and USDA Biobased Product.
  • Compatible with ES10 automatic hand sanitizer dispensers.