Learn More
Cole-Parmer 2196-16-PE Homogenizer Foam Holder, 16 x 50 mL Tubes; 1/EA
Catalog No. NC2115622 Shop All Cole-Parmer Products
Encompass_Preferred
Vial holders are suitable for use with the Spex SamplePrep 2010 Geno/Grinder Homogenizers. Order homogenizer and vials separately.
This item is not returnable. View return policy
Catalog No. NC2115622 Supplier Cole-Parmer Supplier No. 0450027
Vial holders are suitable for use with the Spex SamplePrep 2010 Geno/Grinder Homogenizers. Order homogenizer and vials separately.