Learn More
Research Products International Corp Foam Insert for Vortex-Genie Mixers, Holds 29 - 37 mm Tubes, 2 per Package
SDP
Catalog No. 501978463
Encompass
Two high-density Foam Inserts to accommodate 29 - 37 mm diameter tubes. The insert fits into the Recessed Platform for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
Catalog No. 50-197-8463 Supplier Research Products International Corp Supplier No. 155315
Two high-density Foam Inserts to accommodate 29 - 37 mm diameter tubes. The insert fits into the Recessed Platform for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.