Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer Foam Laboratory Brush, 9"L Handle, 2.5" Diameter; 1/Pk

Catalog No. NC0339033 Shop All Cole-Parmer Products
Encompass_Preferred

Foam Laboratory Washing Brush, 9 in.L

Catalog No. NC0339033 Supplier Cole-Parmer Supplier No. 8455204
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Foam Laboratory Washing Brush, 9 in.L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.