Learn More
Cole-Parmer Foam Universal Glassware Brush, 15"L Handle, 0.75" Diameter; 1/Pk
Catalog No. NC0245626 Shop All Cole-Parmer Products
Encompass_Preferred
Long-lasting brushes last up to 20 times longer than sponges! Soft, plush foam gently cleans interior surfaces without absorbing water -- foam bristles never become soggy or limp! Fits glassware with a 2-"W or larger neck.The universal brush measures " in diameter and bends to conform to the shape of the glassware -- when bent and rotated it cleans corners of the glass that would otherwise be unreachable. Dimensions 16.5 in L x 3 in W x 1.5 in H
Catalog No. NC0245626 Supplier Cole-Parmer Supplier No. 8455202
Long-lasting brushes last up to 20 times longer than sponges! Soft, plush foam gently cleans interior surfaces without absorbing water -- foam bristles never become soggy or limp! Fits glassware with a 2-"W or larger neck.The universal brush measures " in diameter and bends to conform to the shape of the glassware -- when bent and rotated it cleans corners of the glass that would otherwise be unreachable. Dimensions 16.5 in L x 3 in W x 1.5 in H