Learn More
Chemglass Life Sciences Foam Plug, fits 35-45mm Openings
SDP
Catalog No. 501897555
Encompass
Foam Plug, fits 35-45mm Openings. Identi-Plugs are for use with test tubes and flasks. They are reusable and leave no clinging fibers like cotton plugs. They provide a snug fit, yet permit free air flow. Plugs are ideal for use with Drosophila vials. They spring back to original shape after repeated use. Identi-Plugs can also be used as a cleaner for lab pipettes, tubes, vials, as well as industrial precision tubing. Plugs are approximately 1.75" long.
Catalog No. 50-189-7555 Supplier Chemglass Life Sciences Supplier No. CGE1490045
Foam Plug, fits 35-45mm Openings. Identi-Plugs are for use with test tubes and flasks. They are reusable and leave no clinging fibers like cotton plugs. They provide a snug fit, yet permit free air flow. Plugs are ideal for use with Drosophila vials. They spring back to original shape after repeated use. Identi-Plugs can also be used as a cleaner for lab pipettes, tubes, vials, as well as industrial precision tubing. Plugs are approximately 1.75" long.