Learn More
Thomas Scientific Foam Plug for Drosophila Bottles, Blue
SDP
Catalog No. 501461865
Encompass
Our autoclavable Foam Plugs fit both wide and narrow vials, as well as our drosophila bottles. Available in our signature Azer blue.
Catalog No. 50-146-1865 Supplier Thomas Scientific Supplier No. 1182X83
Our autoclavable Foam Plugs fit both wide and narrow vials, as well as our drosophila bottles. Available in our signature Azer blue.