Learn More
Grainger Best Sanitizers, Inc. Foam Pump Kit, Plastic, Clear, 1 ea
Catalog No. 502783114
Encompass_Preferred
This clear plastic foam pump is designed to provide efficient soap dispensing in food processing and food service environments. It is specifically compatible with the MD10201 dispensing system to ensure reliable performance and hygiene.
  • Compatible with MD10201 dispensing systems
  • Constructed from clear, durable plastic
  • Engineered for food processing and food service applications
  • Provides rich, thick foam dispensing
Catalog No. 50-278-3114 Supplier Grainger Supplier No. 19RC72
This clear plastic foam pump is designed to provide efficient soap dispensing in food processing and food service environments. It is specifically compatible with the MD10201 dispensing system to ensure reliable performance and hygiene.
  • Compatible with MD10201 dispensing systems
  • Constructed from clear, durable plastic
  • Engineered for food processing and food service applications
  • Provides rich, thick foam dispensing