Learn More
World Precision Instrument Foam Ring for Glass Storage Jars-1965
SDP
Catalog No. 502521499
Encompass
The foam ring fits inside the storage jars to hold up to 30 of your micropipettes.
Catalog No. 50-252-1499 Supplier World Precision Instrument Supplier No. 1965
The foam ring fits inside the storage jars to hold up to 30 of your micropipettes.