Learn More
Mitegen Foam Ring for Vitrification Dewar By Nanosoft, Compatible with Nanosoft & Vitrobot Dewars
SDP
Catalog No. 5000495023
Encompass
The Nanosoft Foam Ring is a compatible and essential accessory for vitrification dewars. It provides a stable and secure platform for your cryo-EM sample preparation, helping to maintain a controlled environment around your vitrified grids. This foam ring is specifically designed to be compatible with both the NanoSoft Vitrification Dewar and Thermo Fisher Scientific’s/FEI’s Vitrobot Dewar, offering crucial versatility for various lab setups. Its insulating foam construction acts as a thermal barrier, helping to maintain a stable temperature and ensuring your samples remain in optimal condition for imaging. It provides a secure base for handling samples, minimizing the risk of accidental disturbance, and is a simple yet effective tool for improving the vitrification process.
Catalog No. 50-004-95023 Supplier Mitegen Supplier No. MCEMNSVDFR
The Nanosoft Foam Ring is a compatible and essential accessory for vitrification dewars. It provides a stable and secure platform for your cryo-EM sample preparation, helping to maintain a controlled environment around your vitrified grids. This foam ring is specifically designed to be compatible with both the NanoSoft Vitrification Dewar and Thermo Fisher Scientific’s/FEI’s Vitrobot Dewar, offering crucial versatility for various lab setups. Its insulating foam construction acts as a thermal barrier, helping to maintain a stable temperature and ensuring your samples remain in optimal condition for imaging. It provides a secure base for handling samples, minimizing the risk of accidental disturbance, and is a simple yet effective tool for improving the vitrification process.