Learn More
Grainger TapeCase Sealing Foam Tape, Silicone Rubber Sponge, 62.5 mil, 10 yd, 1 ea
Catalog No. 502755092
Encompass_Preferred
This continuous, single-sided sealing tape consists of a conformable polyester foam backing coated with a permanent silicone adhesive. Designed for effective gasketing and sealing, the foam construction provides insulation, dampens sound, and absorbs vibration. It is suitable for both indoor and outdoor use across a wide range of surface materials.
  • 62.5 mil thickness and 3/4 in. width
  • 10 yd. roll length
  • Temperature performance range of -100°F to 500°F
  • 65 lb./in. tensile strength
  • 15 oz./in. adhesion to steel
  • Closed foam cell type with moisture and humidity resistance
  • Meets FDA 21 CFR 177.2605 standards
  • Compatible with diverse surfaces including metal, composites, and various plastics
Catalog No. 50-275-5092 Supplier Grainger Supplier No. 15D298
This continuous, single-sided sealing tape consists of a conformable polyester foam backing coated with a permanent silicone adhesive. Designed for effective gasketing and sealing, the foam construction provides insulation, dampens sound, and absorbs vibration. It is suitable for both indoor and outdoor use across a wide range of surface materials.
  • 62.5 mil thickness and 3/4 in. width
  • 10 yd. roll length
  • Temperature performance range of -100°F to 500°F
  • 65 lb./in. tensile strength
  • 15 oz./in. adhesion to steel
  • Closed foam cell type with moisture and humidity resistance
  • Meets FDA 21 CFR 177.2605 standards
  • Compatible with diverse surfaces including metal, composites, and various plastics