Learn More
Electron Microscopy Sciences Foam Tipped Applicators, Wood Shaft
SDP
Catalog No. 50949149
Encompass_Preferred
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. Head size: 5⁄8"Dia x 1"L
Catalog No. 50-949-149 Supplier Electron Microscopy Sciences Supplier No. 72317
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. Head size: 5⁄8"Dia x 1"L