Learn More
Electron Microscopy Sciences Foam Tipped Applicators, Arrow Tip
SDP
Catalog No. 50949155
Encompass_Preferred
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. 6"polypropylene shaft, arrow end tip 5⁄8"Dia x 7⁄8"L
Catalog No. 50-949-155 Supplier Electron Microscopy Sciences Supplier No. 72315
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. 6"polypropylene shaft, arrow end tip 5⁄8"Dia x 7⁄8"L