Learn More
Cole-Parmer Foam-Tipped Long Glassware Brush, 20" L x 2" Diameter; 1/Pk
Catalog No. NC0497983 Shop All Cole-Parmer Products
Encompass_Preferred
Ergonomically designed wide, flat handles makes for easy rotating of brush. Excellent for all wide mouthed bottles, culture, volumetric graduated, filtering and many other kinds of long flasks, jugs and cylinders. Brushes are tough, yet non abrasive, and dishwasher safe. It works equally well on both plastic and glass bottles. Use with hot water. No detergents needed.
Catalog No. NC0497983 Supplier Cole-Parmer Supplier No. 8455216
Ergonomically designed wide, flat handles makes for easy rotating of brush. Excellent for all wide mouthed bottles, culture, volumetric graduated, filtering and many other kinds of long flasks, jugs and cylinders. Brushes are tough, yet non abrasive, and dishwasher safe. It works equally well on both plastic and glass bottles. Use with hot water. No detergents needed.