Learn More
MSC Rochester Midland Corporation HydroFoamer, 96 oz, Plastic Tank
Catalog No. 502838251
Encompass
The Rochester Midland Corporation HydroFoamer eliminates the need for hand mixing concentrated chemicals with water. Users can add a concentrated solution to the 96 oz tank and connect it to a water source to produce a thick foam spray. Chemicals and water are mixed only during spraying, which helps reduce product waste, and it requires no electricity or complex installation.
  • Built with trusted components, proven reliable through years of field use.
  • Compact and easy to store.
  • No mixing, leading to less wasted time and increased employee productivity.
  • Enhanced safety due to reduced exposure to concentrates.
  • Non-electric, no installation required-just fill the concentrate container and connect to a water source.
  • Maximum cleaning product performance.
Catalog No. 50-283-8251 Supplier MSC Supplier No. 54714209
The Rochester Midland Corporation HydroFoamer eliminates the need for hand mixing concentrated chemicals with water. Users can add a concentrated solution to the 96 oz tank and connect it to a water source to produce a thick foam spray. Chemicals and water are mixed only during spraying, which helps reduce product waste, and it requires no electricity or complex installation.
  • Built with trusted components, proven reliable through years of field use.
  • Compact and easy to store.
  • No mixing, leading to less wasted time and increased employee productivity.
  • Enhanced safety due to reduced exposure to concentrates.
  • Non-electric, no installation required-just fill the concentrate container and connect to a water source.
  • Maximum cleaning product performance.