Learn More
Electron Microscopy Sciences Polyurethane Foam General Purpose Swabs with 2.7mm Pointed Tips
SDP
Catalog No. 501930638
Encompass_Preferred
Economical general purpose swabs. Best solvent retention. The most cost-effective swabs for general purpose applications. 100 ppi reticulated medical grade foam provides excellent particle entrapment. The open cell structure gives the best solvent holding ability of any style of swab. These swabs are free of adhesives or binders that could contaminate your process. Fiberless construction does not generate particles.
Catalog No. 50-193-0638 Supplier Electron Microscopy Sciences Supplier No. 7832583
Add to Cart
Add to Cart
Economical general purpose swabs. Best solvent retention. The most cost-effective swabs for general purpose applications. 100 ppi reticulated medical grade foam provides excellent particle entrapment. The open cell structure gives the best solvent holding ability of any style of swab. These swabs are free of adhesives or binders that could contaminate your process. Fiberless construction does not generate particles.