Description
Effectively PM the applied materials XGEN CVD Poly chamber in a timely manner while maintaining the wafer thiness, the wafer Rs non-uniformities withing specifications and improving tool performance
Specifications
Specifications
| For Use With (Application) | For cleaning critical surfaces |
| Grit | 180, 800, 1350 Grit Diamond |
| Includes | Two 800 Grit Diamond ScrubPAD, 180 Grit Diamond ScrubPAD, 800 Grit Diamond ScrubTIP™, 180 Grit Diamond ScrubTIP™, 1350 Grit Diamond ScrubTIP™, UltraSOLV™ spong |