Learn More
Description
- Protects wearer from contact and provides fresh, clean barrier to laundered mask in hood
- Fiber free
- Ideal for use in strenuous production and maintenance procedures
- Absorbs greater particle control
- Offers longer life and speech clarity
- No restriction of breath
- Avoids moisture build-up on face
Specifications
Specifications
| Size | Universal |
| Material | Foam |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Production, Maintenance |
| Product Type | Cleanroom Beard Cover |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
