Learn More
Foamtec ErgoSCRUB™ Scrubbing Tool
Catalog No. 191202177 Shop All Foamtec International Products
Click to view available options
Description:
Hard Handle
Soft Handle
Soft ergonomic scrubbing tool for implant tools, ceramic domes, Novellus heater blocks, and large vacuum chambers
  • Rounded, ergonomically designed scrubbing tool reduces hand stress and makes cleanroom scrubbing easier
  • Flexible soft handle for use on implant tools, heater blocks, vacuum chambers, and ceramic domes
  • Firmer hard handle provides uniform scrubbing action on flat surfaces, minimizng tool wear
  • Both accommodate a variety of diamond or silicone carbide ScrubDISK™ pads that attach with either pressure-sensitive adhesive or loop backing on reusable styles
Order Info
Available as a reusable or disposable tool. Contact your Fisher Scientific™ Customer Service Representative for details about ScrubDISK options.
Specifications
Product Type Scrubbing Tool
Description Soft Handle
For Use With (Application) For cleaning critical surfaces
For Use With (Equipment) Wet cleans on CVD, PECVD, PVD, Sputtering, Plasma Etch, RIE and Ion Implant Processes in Semiconductor wafer fabs; Domes
Includes Loop backing
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.