Learn More
Foamtec ErgoWRENCH™ Vacuum Chamber Scrubbing Tools: Foam Grip
Catalog No. 19304789 Shop All Foamtec International Products
Change view
Click to view available options
Foam Grip
Catalog No.
19-304-789 Foam Grip
1 options
This item is not returnable. View return policy
Catalog No. 19-304-789 Supplier Foamtec Supplier No. HT5004
This item is not returnable. View return policy
Foam grip for use with ErgoWRENCH Vacuum Chamber Scrubbing Tool
Soft ergonomic grip provides ease of use.
Specifications
Product Type Foam Grip
For Use With (Application) For safer and more efficient scrubbing of hard to reach areas
For Use With (Equipment) All Ergowrenches™ and vat valve heads
Includes Hook
Material Delrin™ Plastic