Learn More
Foamtec Cleanroom Foam Face Mask
Catalog No. 19062636 Shop All Foamtec International Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
19-062636 Single-use
1 options
This item is not returnable. View return policy
Catalog No. 19-062636 Supplier Foamtec Supplier No. HT4401
  • $8.84
    $1.72 / Pack of 10
    Save $7.12
    81% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,769.00
    $430.00 / Case of 250 PK
    Save $1,339.00
    76% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fiber-free cleanroom foam masks provide continuous particle/moisture control while providing cool and flexible comfort.
Provide continuous particle/moisture control and cool, flexible comfort without the speech constraints, goggle/glasses steaming or moisture build-up on the face typical of paper and dense cloth masks
  • Fiber-free
  • Restricts vapor transfer from breath to work surface
  • Breath and speech clarity
  • Reduces lifting or pulling down masks
  • Will not fog safety goggles
  • More absorbent for greater particle control and longer life
Specifications
Product Type Cleanroom Face Mask
Fastening Type Earloops
Material Polyurethane Foam
Disposable Single-use
Includes Ear loops
Model HT4401
Pore Size >0.5 μm