Learn More
Foamtec PharmaMOP™ Cleanroom Mops: Starter Kit
Catalog No. 19304759 Shop All Foamtec International Products
Catalog No. 19-304-759 Supplier Foamtec Supplier No. FS4500PHM
Add to Cart
Add to Cart
For critical cleanroom surfaces allowing the operator to clean ceilings, walls and floors with ease
PharmaMOP utilizes simple press fit connection that enables gloved operators to easily attach mop heads, and is compatible with steam sterilization cycles. Mop heads enable the quick, efficient cleaning of surfaces stained with disinfectant residues
  • Two direction 360° swivel and mop head
  • Abrasion resistant cleanroom fabric
  • Class 100 processing
  • Fiber-free mopping of rough, non-skid floors
  • Sterile
  • Ultra-lightweight telescoping handle
Certifications
ISO 9001:2000 & 13485:2003 certified
Specifications
Product Type Starter Kit
Includes Polyester mop head with foam core: 0.125 in., 0.25 in., EzGLIDE polyester mop head with 0.24 in. foam core, EzGLIDE microfiber mop head; 14 in. mop with locking head frame