Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foamtec PharmaMOP™ SingleSTEP Sahara™ Mop Head

Catalog No. 19304840 Shop All Foamtec International Products
Change view
Click to view available options
Size:
0.25 x 5 x 16 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-304-840 0.25 x 5 x 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-304-840 Supplier Foamtec Supplier No. FS859SH
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Designed for weekly cleaning, this mop strips heavy soils and residues from floors, walls, stainless steel, and windows while allowing even application of disinfectants

  • Designed for walls, floors, and ceilings in ISO Class 1-7 clean rooms
  • Ultra lightweight
  • Two axis swivel reduces work load
  • Individually packed

Specifications

Color White, Green (Foam Strips)
Size 0.25 x 5 x 16 in.
For Use With (Application) Designed for walls, floors, and ceilings in ISO Class 1-7 clean rooms
Material Microfiber Polyester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.