Learn More
Foamtec ScrubCLEAN™ Abrasive Sponges
Catalog No. 19304736 Shop All Foamtec International Products
Change view
Click to view available options
For Use With (Application):
Ink, paste, Epoxy removal from stencils and scrubbing bioreactors
Catalog No. For Use With (Application)
19-304-736 Ink, paste, Epoxy removal from stencils and scrubbing bioreactors
1 options
Catalog No. 19-304-736 Supplier Foamtec Supplier No. FS206
Add to Cart
Add to Cart
For cleaning hardened residues off scratch sensitive tools and components
For components such as CMP polishers, shower heads, heater surfaces, metal/polyester mesh masks, sealing surfaces, plastic and glass surfaces
  • Made of plastic particles that are adhered to sponge with polyurethane polymer
  • Minimizes particle generation from brasive surface
  • Low in endo-toxins, metals, salts and sodium
  • Autoclavable
Specifications
For Use With (Application) Ink, paste, Epoxy removal from stencils and scrubbing bioreactors
Material Polyurethane Polymer