Learn More
Foamtec ScrubWRIGHT™ Chamber Cleaning Tool
Catalog No. 19304733 Shop All Foamtec International Products
Change view
Click to view available options
Grit:
1350
280
360
800
Not Applicable
Product Type:
Scrub Belt
Scrub Pen
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grit Product Type
19-304-733 360 Scrub Belt
19-304-734 800 Scrub Belt
19-304-735 1350 Scrub Belt
19-304-776 Not Applicable Scrub Pen
19-304-732 280 Scrub Belt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-304-733 Supplier Foamtec Supplier No. HT4536DW5
Add to Cart
Add to Cart
This item is not returnable. View return policy
For cleaning small, hard to reach areas
  • Removes deposition from areas critical to process
  • Prevents unwanted flaking
ScrubWRIGHT FTPEN-1
  • High impact plastic w/spring-tensioned attachment mechanism
ScrubBELTS
  • Interchangeable belt-shaped
  • Spring adjustable for easy mounting and rotation
  • Minimizes tool wear
  • For polishing stains, scratches and process-induced residue from scratch sensitive surfaces
Specifications
Product Type Scrub Belt
For Use With (Application) For quick removal of deposition from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces
Grit 360
Material Diamond