Learn More
Foamtec Sterile UltraMOP™ Foam Mop Heads
Catalog No. 19304575 Shop All Foamtec International Products
Change view
Click to view available options
UltraMOP; Perfex style; L x W: 16 x 5 in.;
Catalog No.
19-304-575 UltraMOP; Perfex style; L x W: 16 x 5 in.;
1 options
Catalog No. 19-304-575 Supplier Foamtec Supplier No. FS500ST
Add to Cart
Add to Cart
For wet mopping procedures in sterile and controlled environments.
  • Compatible with Perfex™ TruCLEAN™ mopping system
  • Open cell polyurethane foam construction
  • Gamma irradiated
  • Absorbs spills and applies even volumes of disinfectants