Learn More
Foamtec UltraSOLV™ ScrubDISKs
Catalog No. 19304801 Shop All Foamtec International Products
Change view
Click to view available options
Grit:
140
180
220
280
360
800
1350
2000
3000
4000
Includes:
Hook (loop on handle)
Hook and loop fastener
Hook and loop fastener on handle
Pressure-sensitive adhesive
Pressure-sensitive adhesive or hook and loop fastener
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grit Includes
19-304-801 220 Pressure-sensitive adhesive
19-042-471 180 Hook and loop fastener
19-120-2182 280 Hook and loop fastener
19-120-2192 800 Pressure-sensitive adhesive or hook and loop fastener
19-304-799 140 Pressure-sensitive adhesive
19-304-800 180 Pressure-sensitive adhesive
19-304-802 280 Pressure-sensitive adhesive
19-304-803 1350 Pressure-sensitive adhesive
19-304-804 2000 Pressure-sensitive adhesive
19-304-805 3000 Pressure-sensitive adhesive
19-304-810 3000 Hook and loop fastener on handle
19-304-809 2000 Hook and loop fastener
19-304-806 4000 Pressure-sensitive adhesive
19-304-807 360 Hook (loop on handle)
19-304-811 4000 Hook and loop fastener
19-304-851 1350 Hook and loop fastener
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19-304-801 Supplier Foamtec Supplier No. HT4522DC3G1
Add to Cart
Add to Cart
Request a Quote
Wide grit selection allows for quick removal of process-induced residue from aluminum, stainless steel, ceramic, glass, quartz and anodized surfaces and minimizes tool wear.
  • Patented scrubbing fiber free tool
  • Fiber-free construction for reducing particle levels in cleaned vacuum chambers
  • Unique bonding technology for minimizing release of abrasive particles
  • Eliminates H2O2 from PMs
  • Removes hardened process residue from Hi vacuum chambers
  • For use in conjunction with ErgoSCRUB™ hand tool
Specifications
Product Type Scrub Disk
Description Fiber-free construction
Diameter (English) 3.5 in.
Diameter (Metric) 8.89 cm
Disposable Single-use
For Use With (Application) Quick removal of process induced residue from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces while minimizing tool wear
For Use With (Equipment) ErgoScrub hand tool
Grit 220
Includes Pressure-sensitive adhesive
Material Diamond