Learn More
Medchemexpress LLC Fobrepodacin disodium | 1384984-20-6 | 99.5% | 552.40 g·mol-1 | C21H24FN6Na2O6P | 5 MG
SDP
Catalog No. 5000222501
Encompass_Preferred
Fobrepodacin disodium is the disodium phosphate salt form of an orally active prodrug developed as a gyrase B inhibitor with potent bactericidal activity against mycobacterial pathogens. Supplied as a white to off-white solid for research use, the material is characterized for purity, solubility, and storage to support in vitro and in vivo studies.
  • Orally active phosphate prodrug
  • Potent bactericidal activity in vivo
  • High purity: 99.5%
  • White to off-white solid; hygroscopic-store sealed
  • Soluble in DMSO (~5 mg/mL) with pH adjustment recommended
  • Stable in solvent: -80°C (6 months), -20°C (1 month)
Catalog No. 50-002-22501 Supplier Medchemexpress LLC Supplier No. HY1356555MG
Fobrepodacin disodium is the disodium phosphate salt form of an orally active prodrug developed as a gyrase B inhibitor with potent bactericidal activity against mycobacterial pathogens. Supplied as a white to off-white solid for research use, the material is characterized for purity, solubility, and storage to support in vitro and in vivo studies.
  • Orally active phosphate prodrug
  • Potent bactericidal activity in vivo
  • High purity: 99.5%
  • White to off-white solid; hygroscopic-store sealed
  • Soluble in DMSO (~5 mg/mL) with pH adjustment recommended
  • Stable in solvent: -80°C (6 months), -20°C (1 month)