Learn More
Medchemexpress LLC Fobrepodacin disodium | 1384984-20-6 | 99.5% | C21H24FN6Na2O6P | 10MG
SDP
Catalog No. 5000259991
Encompass_Preferred
Fobrepodacin disodium is the disodium phosphate prodrug of an aminobenzimidazole gyrase B/ParE inhibitor. It is an orally bioavailable antibacterial agent that demonstrates potent bactericidal activity in preclinical in vivo models, including mycobacterial infection studies. Supplied as a white to off-white solid, it is intended for research use in drug discovery and efficacy experiments.
  • Orally bioavailable phosphate prodrug
  • Potent bactericidal activity in vivo
  • Suitable for preclinical efficacy and pharmacology studies
  • White to off-white solid, convenient for formulation
  • High purity (99.5%) for reproducible results
Catalog No. 50-002-59991 Supplier Medchemexpress LLC Supplier No. HY13565510MG
Fobrepodacin disodium is the disodium phosphate prodrug of an aminobenzimidazole gyrase B/ParE inhibitor. It is an orally bioavailable antibacterial agent that demonstrates potent bactericidal activity in preclinical in vivo models, including mycobacterial infection studies. Supplied as a white to off-white solid, it is intended for research use in drug discovery and efficacy experiments.
  • Orally bioavailable phosphate prodrug
  • Potent bactericidal activity in vivo
  • Suitable for preclinical efficacy and pharmacology studies
  • White to off-white solid, convenient for formulation
  • High purity (99.5%) for reproducible results