Learn More
Bioworld Focus/extraction Buffer-I, 50 mL
Supplier:  Bioworld 220400371
Encompass_Preferred
FOCUS™ Extraction Buffer I A modified well studied protein stabilization and IPF strip re-hydration buffer designed to produce better spots on 2D gel anaysis and avoid streacking of membrane proteins. Features of the FOCUS™ Extraction BuffersConvenient and simple to use extraction buffers.No weighing and preparation required, simply hydrate and use.Fresh and ready to use reagents every time.No worries abut urea induced protein carbamylation or waste of unused reagents.No ghost protein bands due to dust and human skin contamination. Technical Information:This buffer comes in a 50 ml bottle and contians Urea and NP-40.
Catalog No. 50-253-036