Learn More
World Precision Instrument Focus Knob Cover, Silicone
SDP
Catalog No. 502539229
Encompass
Silicone handle covers for use with the SurgiScope Surgical Microscope. The silicone handle covers protect the handles and knobs of your microscope during surgery and can be easily removed for cleaning.
Catalog No. 50-253-9229 Supplier World Precision Instrument Supplier No. 505129
Silicone handle covers for use with the SurgiScope Surgical Microscope. The silicone handle covers protect the handles and knobs of your microscope during surgery and can be easily removed for cleaning.