Learn More
Sklar Instruments Foerster Sponge Forceps, Sklar, 9-1/2 in., Curved, Serrated, Stainless Steel. EA/1.
SDP
Catalog No. 501436514 Shop All Sklar Instruments Products
Encompass_Preferred
Foerster Sponge Forceps, Sklar, 9-1/2 in., Curved, Serrated, Stainless Steel. Sklar Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster sponge forceps are often used in bariatric surgeries because of their longer length, and the curved version may be used for removing the placenta in OB/GYN procedures. This product is curved with oval, fenestrated, serrated tips and a length of 9-1/2 inches.
This item is not returnable. View return policy
Catalog No. 50-143-6514 Supplier Sklar Instruments Supplier No. 872395
Foerster Sponge Forceps, Sklar, 9-1/2 in., Curved, Serrated, Stainless Steel. Sklar Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster sponge forceps are often used in bariatric surgeries because of their longer length, and the curved version may be used for removing the placenta in OB/GYN procedures. This product is curved with oval, fenestrated, serrated tips and a length of 9-1/2 inches.